Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Di-tert-amylphenol 98.0+%, TCI America™
SDP

Catalog No. D5320400G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5320400G 400 g
D532025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5320400G Supplier TCI America Supplier No. D5320400G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-95-6
Molecular Formula C16H26O
Molecular Weight (g/mol) 234.38
MDL Number MFCD00041929
InChI Key WMVJWKURWRGJCI-UHFFFAOYSA-N
Synonym 2,4-di-tert-pentylphenol, 2,4-di-tert-amylphenol, 2,4-di-t-pentylphenol, 2,4-bis 1,1-dimethylpropyl phenol, phenol, 2,4-bis 1,1-dimethylpropyl, di-tert-amylphenol, prodox 156, phenol, 2,4-di-tert-pentyl, 2,4-bis 2-methylbutan-2-yl phenol, unii-852f1hp88h
PubChem CID 8455
IUPAC Name 2,4-bis(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)CC

Specifications

Color White-Yellow
Boiling Point 143°C
Quantity 400 g
UN Number 3077
Formula Weight 234.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Di-tert-amylphenol
EINECSNumber (3)-0521&(3)-0526
RTECSNumber SL3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.