CAS RN 12002-99-2
IUPAC Name:
(argentiotellanyl)silver
Synonyms:
silver telluride
Molecular Weight (g/mol):
343.34
Molecular Formula:
Ag2Te
InChi Key:
YRXWPCFZBSHSAU-UHFFFAOYSA-N
SMILES:
[Ag][Te][Ag]
1
–
1
of
1
results
1
Sigma Aldrich Silver(I) telluride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.