accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiOAlO

CAS RN 12003-67-7

IUPAC Name: [(oxidoalumanylidene)oxo]lithiumylium
Synonyms: lithium aluminate
Molecular Weight (g/mol): 65.92
Molecular Formula: AlLiO2
InChi Key: YQNQTEBHHUSESQ-UHFFFAOYSA-N
SMILES: [Li+].[O-][Al]=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)