Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Aluminate, MP Biomedicals

Catalog No. ICN209344 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209344 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209344 Supplier MP Biomedicals, Inc Supplier No. 0520934480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12003-67-7
Molecular Formula AlLiO2
Molecular Weight (g/mol) 65.92
InChI Key YQNQTEBHHUSESQ-UHFFFAOYSA-N
Synonym lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
PubChem CID 123268
IUPAC Name lithium;oxido(oxo)alumane
SMILES [Li+].[O-][Al]=O

Specifications

Quantity 100 g
Formula Weight 65.92
Chemical Name or Material Lithium Aluminate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.