accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOAlY

CAS RN 12005-21-9

IUPAC Name: aluminium(3+) yttrium(3+) trioxidandiide
Synonyms: aluminium(3+) yttrium(3+) trioxidandiide
Molecular Weight (g/mol): 163.88
Molecular Formula: AlO3Y
InChi Key: JNDMLEXHDPKVFC-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Al+3].[Y+3]
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)