Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium aluminum oxide, 99.99% (metals basis)

Catalog No. AA3624122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3624122 100 g
AA3624114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3624122 Supplier Thermo Scientific Chemicals Supplier No. 03624122
Add to Cart

missing translation for 'validMainframeMsg'

Can be used as a sputtering target to prepare bubble memory devices | CAS: 12005-21-9

Can be used as a sputtering target to prepare bubble memory devices.Yttrium aluminum oxide finds application in ceramic, glass and optics due to its thermal stability and insolubility. It is used in the preparation of sputtering bonded targets in chemical processes viz. chemical vapor deposition, atomic thermal and electron beam evaporation. It is also used in solar, water treatment and fuel cell. Further, it is used as coloring and adhesive agents in enamels manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-21-9
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG

Specifications

Melting Point 1760°C
Physical Form Fused Lump
Quantity 100 g
Refractive Index 1.833
Density 4.65 g/mL
Solubility Information Insoluble in water.
Formula Weight 593.62
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Yttrium aluminum oxide
EINECSNumber 234-465-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.