accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12006-85-8

IUPAC Name: boron diiron
Synonyms: boron diiron
Molecular Weight (g/mol): 122.5
Molecular Formula: BFe2
InChi Key: FSDZRQFSRALZQJ-UHFFFAOYSA-N
SMILES: [B].[Fe].[Fe]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results