accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12006-99-4

IUPAC Name: dimolybdenumynylidyneborane
Synonyms: dimolybdenumynylidyneborane
Molecular Weight (g/mol): 202.71
Molecular Formula: BMo2
InChi Key: QROVMOADHGTXCA-UHFFFAOYSA-N
SMILES: B#[Mo]#[Mo]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results