Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Molybdenum boride, 99.5% (metals basis)

Numéro de catalogue. AA4368022
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA43680-22 100 g
AA43680-14 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA43680-22 Fournisseur Thermo Scientific™ Code fournisseur 4368022
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

CAS: 12006-99-4 | BMo2 | 202.71 g/mol

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 12006-99-4
Molecular Formula BMo2
Molecular Weight (g/mol) 202.71
MDL Number MFCD01310400
InChI Key QROVMOADHGTXCA-UHFFFAOYSA-N
IUPAC Name dimolybdenumynylidyneborane
SMILES B#[Mo]#[Mo]

Spécifications

Physical Form Solid
Quantity 100 g
Formula Weight 202.69
ShelfLife 5 Years
EINECSNumber 234-502-8
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.