accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONbOOONbOOBa

CAS RN 12009-14-2

IUPAC Name: barium(2+) diniobiumoylolate
Synonyms: barium(2+) diniobiumoylolate
Molecular Weight (g/mol): 419.13
Molecular Formula: BaNb2O6
InChi Key: FKKSQMDUUDBSGH-UHFFFAOYSA-N
SMILES: [Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results