Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium niobium oxide, 99.9% (metals basis)

Catalog No. AA3913009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3913009 10 g
AA3913018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3913009 Supplier Thermo Scientific Chemicals Supplier No. 03913009
Add to Cart

CAS: 12009-14-2 | BaNb2O6 | 419.13 g/mol

Barium niobium oxide is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12009-14-2
Molecular Formula BaNb2O6
Molecular Weight (g/mol) 419.13
MDL Number MFCD00064751
InChI Key FKKSQMDUUDBSGH-UHFFFAOYSA-N
Synonym barium niobate, barium niobium oxide, barium 2+ ; oxido dioxo niobium, barium-niobium-oxide, barium 2+ diniobiumoylolate
PubChem CID 6093852
IUPAC Name barium(2+) diniobiumoylolate
SMILES [Ba++].[O-][Nb](=O)=O.[O-][Nb](=O)=O

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN1564
Solubility Information Insoluble in water
Formula Weight 419.11
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Barium niobium oxide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 234-543-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.