accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12012-35-0

IUPAC Name: {[(chromiomethyl)chromio]methylidyne}chromium
Synonyms: chromium carbide
Molecular Weight (g/mol): 180.01
Molecular Formula: C2Cr3
InChi Key: UFGZSIPAQKLCGR-UHFFFAOYSA-N
SMILES: [Cr]#C[Cr]C#[Cr]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results