accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120120-26-5

IUPAC Name: triethyl(trifluoromethyl)silane
Synonyms: triethyl(trifluoromethyl)silane
Molecular Weight (g/mol): 184.28
Molecular Formula: C7H15F3Si
InChi Key: ZHSKFONQCREGOG-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)