Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-Tetrahydrofurfurylamine

Catalog No. 4129371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-293-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 41-293-71G Supplier Sigma Aldrich Supplier No. 4129371G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-Tetrahydrofurfurylamine

Specifications

CAS 120120-26-5
Density 0.98 g/mL at 25°C
Boiling Point 56°C to 57°C (60 mmHg, lit.)
Molecular Formula C7H15F3Si
Refractive Index n20/D 1.455
Linear Formula (C2H5)3SiCF3
MDL Number MFCD00192520
Quantity 1 g
Synonym (R)-2-(Aminomethyl)tetrahydrofuran
Molecular Weight (g/mol) 184.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.