CAS RN 12017-12-8
CAS RN 12017-12-8
IUPAC Name:
(silanylidenecobaltio)silane
Synonyms:
(silanylidenecobaltio)silane
Molecular Weight (g/mol):
115.1
Molecular Formula:
CoSi2
InChi Key:
QKWVWIGBHQUEHX-UHFFFAOYSA-N
SMILES:
[Si]=[Co]=[Si]
1
–
1
of
1
results
1
Cobalt silicide, 99% (metals basis)
CAS: 12017-12-8 Molecular Formula: CoSi2 Molecular Weight (g/mol): 115.10 MDL Number: MFCD00016021 InChI Key: QKWVWIGBHQUEHX-UHFFFAOYSA-N