accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120186-59-6

IUPAC Name: [4-(benzyloxy)phenyl](bromo)magnesium
Synonyms: [4-(benzyloxy)phenyl](bromo)magnesium
Molecular Weight (g/mol): 287.44
Molecular Formula: C13H11BrMgO
InChi Key: NMSSOXCJYITCNG-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)