Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexafluorozirconic Acid Solution

Catalog No. 6797551L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-975-51L 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-975-51L Supplier Sigma Aldrich Supplier No. 6797551L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used for:Hexafluorozirconic acid based surface pretreatments on steel for corrosion resistancePreparation of titania photocatalyst synthesized from ionic-liquid-like precursorSynthesis of fluoride-releasing dental monomerAs precursor to ZrO2 ceramic thin films

Specifications

CAS 120186-59-6
Density 1.033 g/mL at 25°C
Molecular Formula C13H11BrMgO
Linear Formula C13H11BrMgO
MDL Number MFCD09039130
Quantity 100 mL
Synonym Dihydrogen hexafluorozirconate solution; Hydrogen zirconium fluoride
Molecular Weight (g/mol) 287.43 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.