accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1202-34-2

IUPAC Name: N-(pyridin-2-yl)pyridin-2-amine
Synonyms: N-2-pyridinyl-2-pyridinamine
Molecular Weight (g/mol): 171.2
Molecular Formula: C10H9N3
InChi Key: HMMPCBAWTWYFLR-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)