Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dipyridylamine, MP Biomedicals

Catalog No. ICN212350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212350 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212350 Supplier MP Biomedicals, Inc Supplier No. 0521235005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metal-complexing agent suitable for research and further manufacturing use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1202-34-2
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.203
InChI Key HMMPCBAWTWYFLR-UHFFFAOYSA-N
Synonym 2,2'-dipyridylamine, di 2-pyridyl amine, di pyridin-2-yl amine, 2,2'-bipyridylamine, 2-pyridinamine, n-2-pyridinyl, n-pyridin-2-yl pyridin-2-amine, unii-x9bf664yak, pyridinamine, n-pyridinyl, pyridine, 2,2'-iminodi, 2,2-dipyridylamine
PubChem CID 14547
IUPAC Name N-pyridin-2-ylpyridin-2-amine
SMILES C1=CC=NC(=C1)NC2=CC=CC=N2

Specifications

Quantity 10 g
Formula Weight 171.2
Percent Purity 99.8%
Grade Not Available
Physical Form Powder or Flakes
Chemical Name or Material 2, 2«-Dipyridylamine
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.