accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12022-71-8

IUPAC Name: dititanium(4+) diiron(3+) heptaoxidandiide
Synonyms: dititanium(4+) diiron(3+) heptaoxidandiide
Molecular Weight (g/mol): 319.42
Molecular Formula: Fe2O7Ti2
InChi Key: JCDAAXRCMMPNBO-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ti+4].[Ti+4].[Fe+3].[Fe+3]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)