accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12024-14-5

IUPAC Name: gallanetellurol
Synonyms: gallium(II) telluride
Molecular Weight (g/mol): 200.35
Molecular Formula: GaH3Te
InChi Key: SKPFTVFWBLFTTD-UHFFFAOYSA-M
SMILES: [GaH2][TeH]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results