Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(II) telluride, 99.999% (metals basis)

Catalog No. AA4188103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4188103 1 g
AA4188106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4188103 Supplier Thermo Scientific Chemicals Supplier No. 04188103
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12024-14-5 | GaTe | 197.323 g/mol

Gallium(II) telluride acts as a semiconductor is used in the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-14-5
Molecular Formula GaTe
Molecular Weight (g/mol) 197.323
MDL Number MFCD00135536
InChI Key OFIYHXOOOISSDN-UHFFFAOYSA-N
Synonym gallium ii telluride
PubChem CID 14496722
IUPAC Name tellanylidenegallium
SMILES [Ga]=[Te]

Specifications

Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 197.32
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Gallium(II) telluride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 234-690-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.