accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120277-69-2

IUPAC Name: 3-bromo-5-(chloromethyl)pyridine
Synonyms: 3-bromo-5-(chloromethyl)pyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: NLPHAZLCNNDGPS-UHFFFAOYSA-N
SMILES: ClCC1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results