accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1202865-03-9

IUPAC Name: 2-[bis(4-methylphenyl)phosphanyl]benzaldehyde
Synonyms: 2-[bis(4-methylphenyl)phosphanyl]benzaldehyde
Molecular Weight (g/mol): 318.36
Molecular Formula: C21H19OP
InChi Key: IDDRQJVTYBKJLP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results