Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Di-p-tolylphosphino)benzaldehyde, 97%, Thermo Scientific™

Catalog No. AAH6318306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6318306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6318306 Supplier Thermo Scientific Chemicals Supplier No. H6318306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1202865-03-9
Molecular Formula C21H19OP
Molecular Weight (g/mol) 318.356
MDL Number MFCD15143661
InChI Key IDDRQJVTYBKJLP-UHFFFAOYSA-N
Synonym 2-di-p-tolylphosphino benzaldehyde, 2-bis 4-methylphenyl phosphanyl benzaldehyde, acmc-20aon9, 2-bis 4-methylphenyl phosphino benzaldehyde, 2-di-p-tolylphosphino benzaldehyde,min
PubChem CID 71310894
IUPAC Name 2-bis(4-methylphenyl)phosphanylbenzaldehyde
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=CC=CC=C3C=O

Specifications

Melting Point 105°C to 111°C
Color Yellow
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 318.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(Di-p-tolylphosphino)benzaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.