accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOKTa

CAS RN 12030-91-0

IUPAC Name: tantalum(5+) potassium trioxidandiide
Synonyms: tantalum(5+) potassium trioxidandiide
Molecular Weight (g/mol): 268.04
Molecular Formula: KO3Ta
InChi Key: HGMNOOPONDJVDV-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[K+].[Ta+5]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results