Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tantalate, MP Biomedicals

Catalog No. ICN225520 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225520 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225520 Supplier MP Biomedicals, Inc Supplier No. 0522552025

Chemical Identifiers

CAS 12030-91-0
Molecular Formula KO3Ta
Molecular Weight (g/mol) 268.043
InChI Key GTFKZOWFWFAWLD-UHFFFAOYSA-N
Synonym potassium oxido dioxo tantalum, potassium ion tantalumoylolate, potassium tantalumoylolate, potassium tantalate,-100 mesh, potassium tantalate, ktao3, potassium tantalum oxide, puratronic
PubChem CID 3523997
IUPAC Name potassium;oxido(dioxo)tantalum
SMILES [O-][Ta](=O)=O.[K+]

Specifications

Quantity 100 g
Formula Weight 268.04
Chemical Name or Material Potassium Tantalate, KTaO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.