accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12031-80-0

IUPAC Name: dilithium(1+) dioxidanediide
Synonyms: lithium peroxide
Molecular Weight (g/mol): 45.88
Molecular Formula: Li2O2
InChi Key: HPGPEWYJWRWDTP-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O-][O-]
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)