Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium peroxide, 95%, Thermo Scientific™

Catalog No. AC389880100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389880100 10 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC389880100 Supplier Thermo Scientific Chemicals Supplier No. 389880100
  • $141.60
    $120.65 / Each of 1

    Save $20.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-80-0
Molecular Formula Li2O2
Molecular Weight (g/mol) 45.88
MDL Number MFCD00016184
InChI Key HPGPEWYJWRWDTP-UHFFFAOYSA-N
Synonym lithium peroxide li2 o2, lithium peroxide un1472 oxidizer, lithium oxide, dilithium peroxide, unii-9anx556r5f
PubChem CID 25489
IUPAC Name dilithium;peroxide
SMILES [Li+].[Li+].[O-][O-]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 95%
Linear Formula LiO2Li
Formula Weight 45.88
Percent Purity 95%
Packaging Plastic Bottle
Chemical Name or Material Lithium peroxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 234-758-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.