accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiLiOOOTi

CAS RN 12031-82-2

IUPAC Name: titanium(4+) dilithium(1+) trioxidandiide
Synonyms: lithium titanate
Molecular Weight (g/mol): 109.74
Molecular Formula: Li2O3Ti
InChi Key: AXQWGBXVDWYWDE-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O--].[O--].[O--].[Ti+4]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)