Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium titanate

Catalog No. 400939100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-093-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-093-9100G Supplier Sigma Aldrich Supplier No. 400939100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Lithium titanate (LTO) can be used as an anode material, which shows an ion conductivity of 103 Scm1 at room temperature. It can also be used as an alternative to conventional graphite materials. LTO can further be used in the fabrication of high-performance lithium-ion batteries for electric vehicles (EVs).

Specifications

CAS 12031-82-2
Quantity 100 g
Molecular Formula Li2O3Ti
Linear Formula Li2TiO3
MDL Number MFCD00016181
Synonym LTO
Molecular Weight (g/mol) 109.75
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.