accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12032-89-2

IUPAC Name: ditellanylidenemanganese
Synonyms: ditellanylidenemanganese
Molecular Weight (g/mol): 310.14
Molecular Formula: MnTe2
InChi Key: KDJKBQYOEWVJGP-UHFFFAOYSA-N
SMILES: [Te]=[Mn]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results