CAS RN 12033-89-5
Silicon(IV) nitride, alpha-phase, ca. 90%
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Silicon(IV) nitride, alpha-phase, 99.9% (metals basis)
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41
Sigma Aldrich Silicon nitride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Silicon nitride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Silicon nitride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Silicon nitride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Silicon Nitride, Electronic Grade, MP Biomedicals
CAS: 12033-89-5 Molecular Formula: N4Si3 Molecular Weight (g/mol): 140.28 MDL Number: MFCD00011230 InChI Key: HQVNEWCFYHHQES-UHFFFAOYSA-N Synonym: silicon nitride,silicon nitride si3n4,unii-qhb8t06idk,qhb8t06idk,silicon iv nitride, alpha phase,trisilicon tetranitride,nierite,nano silicon nitride,silicon iv nitride,silicon nitride, amorphous PubChem CID: 3084099 IUPAC Name: Silicon nitride SMILES: N12[Si]34N5[Si]11N3[Si]25N41