Learn More
Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase
Description
Silicon(IV) nitride is used in automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1900°C (sublimation) |
| Color | Gray to Black |
| Physical Form | Powder |
| Quantity | 250 g |
| Odor | Odorless |
| Density | 3.2 g/mL |
| Linear Formula | Si3N4 |
| Merck Index | 14,8497 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 140.28 |
| Show More |
Safety and Handling
| EINECSNumber | 234-796-8 |
| RTECSNumber | VW0650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.