CAS RN 12036-09-8
IUPAC Name:
dioxorhenium
Synonyms:
rhenium(IV) oxide
Molecular Weight (g/mol):
218.21
Molecular Formula:
O2Re
InChi Key:
WXBOMIKEWRRKBB-UHFFFAOYSA-N
SMILES:
O=[Re]=O
1
–
1
of
1
results
1
Sigma Aldrich Rhenium(IV) oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.