CAS RN 12036-69-0
IUPAC Name:
titanium(4+) dizinc(2+) tetraoxidandiide
Synonyms:
zinc titanate
Molecular Weight (g/mol):
242.62
Molecular Formula:
O4TiZn2
InChi Key:
ZBFOLPMOGPIUGP-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[Ti+4].[Zn++].[Zn++]
1
–
1
of
1
results
1
Promotions Available
Zinc titanium oxide, 99.9% (metals basis)
Zinc titanium oxide, 99.9% (metals basis)
CAS: 12036-69-0 Molecular Formula: O4TiZn2 Molecular Weight (g/mol): 242.62 MDL Number: MFCD00054107 InChI Key: ZBFOLPMOGPIUGP-UHFFFAOYSA-N Synonym: Zinc titanate