Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc titanium oxide, 99.9% (metals basis)

Catalog No. AA4167318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4167318 50 g
AA4167330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4167318 Supplier Thermo Scientific Chemicals Supplier No. 04167318
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12036-69-0 | O4TiZn2 | 242.62 g/mol

Zinc titanium oxide is used as a regenerable catalyst, a pigment and a sorbent of sulfur compounds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-69-0
Molecular Formula O4TiZn2
Molecular Weight (g/mol) 242.62
MDL Number MFCD00054107
InChI Key ZBFOLPMOGPIUGP-UHFFFAOYSA-N
Synonym Zinc titanate

Specifications

Physical Form Powder
Quantity 50 g
Odor Odorless
Solubility Information Insoluble in water.
Formula Weight 242.64
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc titanium oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H332
Harmful if inhaled.
Precautionary Statement GHS P Statement
P261-P271-P304+P340-P312
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.