accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12039-79-1

IUPAC Name: (silanylidenetantalio)silane
Synonyms: (silanylidenetantalio)silane
Molecular Weight (g/mol): 241.15
Molecular Formula: H4Si2Ta
InChi Key: JSCUKKCIAVANLH-UHFFFAOYSA-N
SMILES: [SiH2]=[Ta]=[SiH2]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results