accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12040-02-7

IUPAC Name: stannanetellone
Synonyms: tin telluride
Molecular Weight (g/mol): 248.33
Molecular Formula: H2SnTe
InChi Key: UURRKPRQEQXTBB-UHFFFAOYSA-N
SMILES: [SnH2]=[Te]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results