accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12044-50-7

IUPAC Name: diarsooxidane hydrate
Synonyms: arsenic pentoxide hydrate
Molecular Weight (g/mol): 247.85
Molecular Formula: As2H2O6
InChi Key: XSIWZGKXZLITBO-UHFFFAOYSA-N
SMILES: O.O=[As](=O)O[As](=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results