accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12057-17-9

IUPAC Name: manganese(4+) manganese(3+) lithium(1+) tetraoxidandiide
Synonyms: manganese(4+) manganese(3+) lithium(1+) tetraoxidandiide
Molecular Weight (g/mol): 180.81
Molecular Formula: LiMn2O4
InChi Key: KEZPOJYZIWMPGY-UHFFFAOYSA-N
SMILES: [Li+].[O--].[O--].[O--].[O--].[Mn+3].[Mn+4]
Quantity 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)