Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Manganese Oxide Spinel (lmo) Powder Battery Grade

Catalog No. 91643950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-643-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-643-950G Supplier Sigma Aldrich Supplier No. 91643950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium Manganate (LiMn2O4) is a cathode material with a spinel structure, which allows the material to be discharged at high rates. LMO-based batteries are most suited for use in high rate applications.NANOMYTE is a registered trademark of NEI Corporation

Specifications

Quantity 50 g
Molecular Formula LiMn2O4
Linear Formula LiMn2O4
Synonym LMO; Lithium manganese(III,IV) oxide; NANOMYTE™ BE-30
Molecular Weight (g/mol) 180.81
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.