accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12058-66-1

IUPAC Name: disodium oxostannanebis(olate)
Synonyms: sodium stannate
Molecular Weight (g/mol): 212.69
Molecular Formula: Na2O3Sn
InChi Key: TVQLLNFANZSCGY-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][Sn]([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results