accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12060-34-3

IUPAC Name: pentaniobium trisilicon
Synonyms: pentaniobium trisilicon
Molecular Weight (g/mol): 548.79
Molecular Formula: Nb5Si3
InChi Key: YSJRXBWBYGDQAI-UHFFFAOYSA-N
SMILES: [Si].[Si].[Si].[Nb].[Nb].[Nb].[Nb].[Nb]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results