Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium silicide, 99.5% (metals basis)

Catalog No. AA3947122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3947122 100 g
AA3947114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3947122 Supplier Thermo Scientific Chemicals Supplier No. 03947122
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12060-34-3 | Nb5Si3 | 548.787 g/mol

The Nb-based RMIC’s have much higher potential application temperatures, because of their dramatically higher melting temperatures. It is used in high temperature in situ composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-34-3
Molecular Formula Nb5Si3
Molecular Weight (g/mol) 548.787
MDL Number MFCD00213811
InChI Key YSJRXBWBYGDQAI-UHFFFAOYSA-N
PubChem CID 13848090
IUPAC Name niobium;silicon
SMILES [Si].[Si].[Si].[Nb].[Nb].[Nb].[Nb].[Nb]

Specifications

Physical Form Powder
Quantity 100 g
Appearance Powder
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 548.79
Chemical Name or Material Niobium silicide
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.