accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12063-19-3

IUPAC Name: bis(ironoylol) zinc
Synonyms: bis(iron hydroxide) zinc
Molecular Weight (g/mol): 243.08
Molecular Formula: Fe2H2O4Zn
InChi Key: NNGHIEIYUJKFQS-UHFFFAOYSA-L
SMILES: [Zn].O[Fe]=O.O[Fe]=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)