Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc iron oxide

Catalog No. 63384410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-384-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-384-410G Supplier Sigma Aldrich Supplier No. 63384410G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc iron oxide nanoparticles are chemically stable and exhibit superparamagnetic properties. There are a number of methods to prepare ZnO nanoparticles, some of them are self-propagated combustion of fuels (alanine, glycine and proline), co-precipitation, sol-gel reaction, spin coating, ball milling and auto combustion.Potential applications of ZnFe2O4 are gas sensors, catalysts and MRI contrast agents.Product of Engi-Mat Co.

Specifications

CAS 12063-19-3
Quantity 10 g
Molecular Formula Fe2O4Zn
Linear Formula ZnFe2O4
MDL Number MFCD00049621
Synonym Diiron zinc tetraoxide
Molecular Weight (g/mol) 241.08
Percent Purity >99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.