accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12063-56-8

IUPAC Name: iron(3+) yttrium(3+) trioxidandiide
Synonyms: iron(3+) yttrium(3+) trioxidandiide
Molecular Weight (g/mol): 192.75
Molecular Formula: FeO3Y
InChi Key: SUFJITMMSUGKIE-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Fe+3].[Y+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results