Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron yttrium oxide, 99.9% (REO)

Catalog No. AA3948822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3948822 100 g
AA3948814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3948822 Supplier Thermo Scientific Chemicals Supplier No. 03948822
  • $351.50
    $283.65 / Each of 1

    Save $67.85

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12063-56-8 | Fe5O12Y3 | 737.931 g/mol

Iron yttrium oxide is mainly used in superconductors. It is also used in filters, transmitters, transducers, solid state lasers in faraday rotators and in non-linear optics. It finds application in polymers, textiles, fuel cell layers, composites and solar energy materials, bio nano materials and bio sensors. Further, it is used in the preparation of fuel cells and solar energy materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-56-8
Molecular Formula Fe5O12Y3
Molecular Weight (g/mol) 737.931
MDL Number MFCD00210618
InChI Key UVXIKKWNYGPENJ-UHFFFAOYSA-N
Synonym iron yttrium oxide, yttrium iron garnet, yttrium ferrite garnet, bis ferric oxide ; oxo oxoyttrio oxy iron; oxo oxoyttrio oxy yttrium, yttrium iron oxide, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16217670
IUPAC Name oxo(oxoferriooxy)iron;oxo(oxoferriooxy)yttrium;oxo(oxoyttriooxy)yttrium
SMILES O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Y]=O.O=[Y]O[Y]=O

Specifications

Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 737.94
Odor Odorless
Appearance Green powder
Chemical Name or Material Iron yttrium oxide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-053-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.