accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(R)H3C(S)(R)O(S)H3CNHNONNH3CO

CAS RN 120685-11-2

IUPAC Name: N-[(15R,16S,17R,18S)-17-methoxy-17,18-dimethyl-5-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaen-16-yl]-N-methylbenzamide
Synonyms: midostaurin
Molecular Weight (g/mol): 570.65
Molecular Formula: C35H30N4O4
InChi Key: IIQIEMHSDLLZQA-QZPVEUDVSA-N
SMILES: CO[C@]1(C)[C@@H]([C@H]2O[C@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O)N(C)C(=O)C1=CC=CC=C1
Product Type 
  • (1)
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results